3,3'-Dipropylthiacarbocyanine iodide 98, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

318434-250MG
318434-250MGEA 103 GBP
318434-250MG
3,3'-Dipropylthiacarbocyanine iodide 98, Sigma-Aldrich®
3,3'-Dipropylthiacarbocyanine iodide

3,3'-Dipropylthiacarbocyanine iodide,98% (3-Propyl-2-[3-[3-propyl-2(3H)benzothiazolylidene]-1-propenyl]benzothiazolium iodide; DiSC3(3))

Formula: C₂₃H₂₅IN₂S₂
MW: 520.491 g/mol
Melting Pt: 288 °C (dec., lit.)
Storage Temperature: Ambient
CAS Number: 53336-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44e331d6fd385e2f19aa27323d4ab3d50c9eb4c8&vid=bb8a83b43aa8f475935c86c5991e3ece9c9c0332&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR