Iron(III) bromide, anhydrous 98+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ferric bromide, Iron tribromide

31125.14 31125.22 31125.06
31125.14EA 169 GBP
31125.14 31125.22 31125.06
Iron(III) bromide, anhydrous 98+%
Iron (III) bromide

Purity: 98+% CAS No.: 10031-26-2 Molecular Formula: FeBr3 Formula Weight: 295.57 Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: Dark red Odour: Odourless Melting Point: dec. Merck Reference: 14,4018 MDL No.: MFCD00016082

Formula: Br₃Fe
MW: 295.56 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00016082
CAS Number: 10031-26-2
EINECS: 233-089-1
UN: 3260
ADR: 8 III
Merck Index: 14,4018

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR