Periodic acid 99.0-101.0% ACS

Supplier: Thermo Fisher Scientific
Danger

Synonyms: ortho-Periodic acid, Orthoperiodic acid

30472.18
30472.18EA 116 GBP
30472.18
Periodic acid 99.0-101.0% ACS
Periodic acid

Purity: 99.01-101.0% CAS No.: 10450-60-9 Molecular Formula: H5IO6 Formula Weight: 227.94 Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: White to pale yellow Odour: Odourless pH: 1.2 Melting Point: 126-130° Merck Reference: 14,7171 UN No.: 3085 MDL No.: MFCD00011344

Formula: H₅IO₆
MW: 227.94 g/mol
Melting Pt: 126…130 °C
Density: 1.3875 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1 II
Merck Index: 14,7171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR