Drill head for carcass sampling
Supplier: BURKLE
5388-1010
300-0356EA 159 GBP
300-0356
Drill head for carcass sampling
Tool Sets
The drill head for carcass sampling has a hexagonal socket for bit holders or chucks, suitable for all commonly used cordless screwdrivers or drilling machines.
  • For sampling in accordance with DIN EN ISO 17604:2015-12
  • Cutting surface: 25 mm Ø
Meat sampler for microbiological sampling of carcass surfaces. Slaughtering businesses (slaughterhouses, butcher's shops, etc.) are obligated as part of operational process control and for monitoring of slaughter hygiene to conduct regular microbiological examinations of carcasses for cattle, hogs, sheep, goats,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable