Cheese trier
Supplier: GERBER FUNKE
3122 3120 3121 3124
300-0145EA 204 GBP
300-0145 300-0143 300-0144 GERB3124
Cheese trier
Samplers Solid Samplers
Chrome-nickel steel. Developed for sampling small cores and testing the readiness of cheeses without cutting them open. Ideal for use in milk-processing industries and cheese dairies.
  • Black plastic handle
  • Two different sizes available