Manually operated reels
Supplier: BURKLE
5309-0010 5309-0020 5309-0030
300-1023EA 146 GBP
300-1023 300-0015 300-0056
Manually operated reels
Hand Reels
The manually operated drum reels are used to lower dipping bottles and immersion cylinders safely to the required depth in shafts, wells and tanks and then to pull them up again. The manually operated drum reels can be equipped with various cables or chains. If flammable or highly flammable liquids are to be collected, then electrically conductive manually operated drum reels must be used together with an earthing cable. The dipping bottles must be made of non sparking metal.
  • High winding capacity of up 150 m for cable 2 mm Ø
  • Reel width: Inner 44 mm,
More Product Information