Hook and loop fastener SJ352Bbk
Supplier: 3M
SJ352BBK
293-0004EA 17.9 GBP
293-0004
Hook and loop fastener SJ352Bbk
Adhesives
This high-performance PSA nylon hook and loop with a rubber adhesive and polypropylene film liner. It provides a firm adhesive bond to a wide variety of surfaces, including many low surface energy plastics and powder coats. It's designed for indoor use and has an engaged thickness of 4,4 mm.
  • High-performance rubber based adhesive designed for indoor applications requiring medium temperature resistance
  • Provides a firm adhesive bond to a wide variety of surfaces
  • Woven nylon is durable for thousands of closures
  • Quick fastening helps reduce production time and
More Product Information