Disodium hydrogen phosphate dihydrate 99,5, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals

Synonyms: sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt

28029.292EA 42.7 GBP
28029.292 28029.235 28029.260 28029.320 28029.460
Disodium hydrogen phosphate dihydrate 99,5, AnalaR NORMAPUR® zA
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
MW: 177.99 g/mol
Melting Pt: 92.5 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7
EINECS: 231-448-7

Order Now

Specification Test Results

Assay Min. 99.5 %
Identification Passes test
Solution in water Passes test
pH (20°C; 5 %) 9.0 - 9.2
Heavy metals (as Pb) Max. 10 ppm
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 20 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 0.5 ppm
Cu (Copper) Max. 3 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR