Spanner wrenches
Supplier: Cytiva
28-9647-78 28-9647-76 28-9647-77
28-9647-78EA 103 GBP
28-9647-78 28-9647-76 28-9647-77
Spanner wrenches
Wrenches
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.