Sodium periodate 99, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Sodium metaperiodate

27986.235EA 132 GBP
27986.235 27986.460 27986.290
Sodium periodate 99, GPR RECTAPUR®
Sodium periodate
Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300
Melting Pt: 175
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1 II
Merck Index: 13,08714

Order Now

Specification Test Results

Assay Min. 99 %
Cl + ClO3 + Br + BrO3 (as Cl) Max. 100 ppm
SO4 (Sulphate) Max. 50 ppm
Mn (Manganese) Max. 1 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR