Sodium fluoride 98,5-100,5 Ph. Eur.

Supplier: VWR Chemicals
Danger

27859.293EA 122 GBP
27859.293 27859.464
Sodium fluoride 98,5-100,5 Ph. Eur.
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700
Melting Pt: 993
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on dried substance) 98.5 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Identification C Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
Cl (Chloride) Max. 200 ppm
Fluorosilicates Passes test
SO4 (Sulphate) Max. 200 ppm
Loss on drying (130°C) Max. 0.5 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR