tri-Sodium citrate dihydrate 99,0-101,0, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. zA

Supplier: VWR Chemicals

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

27833.294EA 57.4 GBP
27833.294 27833.363 27833.237 27833.460 27833.260
tri-Sodium citrate dihydrate 99,0-101,0, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. zA
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
REACH: 01-2119457027-40
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) 99.0 - 101.0 %
Acidity or alkalinity Passes test Ph.Eur.
Appearance of solution Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Readily carbonisable substances Passes test Ph.Eur.
Tartaric acid Passes test
pH (25°C; 5 %) 7.5 - 9.0
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Oxalic acid Max. 100 ppm
Total N (Nitrogen) Max. 10 ppm
Water 11.0 - 13.0 %
Cl (Chloride) Max. 10 ppm
NH3 (Ammonia) Max. 30 ppm
PO4 (Phosphate) Max. 20 ppm
SO4 (Sulphate) Max. 40 ppm
As (Arsenic) Max. 1 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Li (Lithium) Max. 5 ppm
Pb (Lead) Max. 5 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph. Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR