Sodium hydrogen carbonate 99, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: Sodium bicarbonate

27776.296EA 32.9 GBP
27776.296 27776.365 27776.460
Sodium hydrogen carbonate 99, GPR RECTAPUR®
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 150 ppm
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 10 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR