Saponin, GPR RECTAPUR®

Supplier: VWR Chemicals

27534.187EA 170 GBP
27534.187
Saponin, GPR RECTAPUR®
Saponin
Density: 1.015…1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2
EINECS: 232-462-6

Order Now

Specification Test Results

Foam test (5 %; water) 2 - 6 cm
Ignition residue (SO4) Max. 10 %
Insolubility in water Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR