Acetic acid 99,8-100,5, Puriss., meetSpec_BP,Ph.Eur.,USP,FCC, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

27225-10KG 27225-1L 27225-2.5L 27225-30KG 27225-4X2.5L 27225-60KG 27225-6X1L
27225-10KGEA 303 GBP
27225-10KG 27225-1L 27225-2.5L 27225-30KG 27225-4X2.5L 27225-60KG 27225-6X1L
Acetic acid 99,8-100,5, Puriss., meetSpec_BP,Ph.Eur.,USP,FCC, Fluka™
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1.013 hPa)
Melting Pt: 16 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
Assay 99.8 - 100.5 %
Congealing - Freezing Point Min. 15.6 °C
Non-volatile matter Max. 0.005 %
Aluminium (Al) Max. 0.00005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 2 ppm
Lead (Pb) Max. 0.5 ppm
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 2 ppm
Chloride (Cl) complying
Sulfate (SO₄) complying
KMnO₄ reducing matter (as HCOOH) Max. 0.01 %
KMnO₄ reducing substances complying
K₂Cr₂O₇ reducing matter complying
Residual solvents complying
Identity USP complying
Explosion Limit 4 %, 59 °F
Vapor Density 2.07 (vs air)
Refractive Index n20/D 1.371 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR