Citric acid, anhydrous 99,5-100,5 Anhydrous basis, meetSpec_BP,Ph.Eur.,USP,E330, Fluka™

Supplier: Honeywell Chemicals
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

27109-1KG 27109-2.5KG 27109-25KG 27109-6X1KG 27109-6X2.5KG 27109-100G
27109-1KGEA 66 GBP
27109-1KG 27109-2.5KG 27109-25KG 27109-6X1KG 27109-6X2.5KG 27109-100G
Citric acid, anhydrous 99,5-100,5 Anhydrous basis, meetSpec_BP,Ph.Eur.,USP,E330, Fluka™
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: 153…159 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on the anhydrous basis) 99.5 - 100.5 %
Water (Karl Fischer) Max. 0.5 %
Sulfated ash Max. 0.05 %
Aluminium (Al) Max. 0.2 ppm
Arsenic (As) Max. 1 ppm
Calcium (Ca) Max. 0.02 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.001 %
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 0.5 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.005 %
Oxalate (as Oxalic acid) Max. 100 ppm
Sulfate (SO₄) Max. 100 ppm
Reaction against H₂SO₄ complying
Residual solvents complying
Appearance of the solution complying
Explosion Limit 8 %, 65 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR