Potassium bromide 99,5, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Warning

26708.238EA 32.1 GBP
26708.238 26708.295
Potassium bromide 99,5, AnalaR NORMAPUR® zA
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Assay Min. 99.5 %
Acidity or alkalinity Passes test
pH (20°C; 5 %) 5 - 8
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Loss on drying (105°C) Max. 0.5 %
BrO3 (Bromate) Max. 10 ppm
Cl (Chloride) Max. 0.1 %
I (Iodide) Max. 10 ppm
IO3 (Iodate) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
Ba (Barium) Max. 20 ppm
Ca (Calcium) Max. 10 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 10 ppm
Na (Sodium) Max. 0.02 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR