Tetrachloroethylene 99, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene

26218.292EA 53.5 GBP
26218.292 26218.326 26218.361 26218.463
Tetrachloroethylene 99, GPR RECTAPUR®
Tetrachloroethylene
Formula: C₂Cl₄
MW: 165.83 g/mol
Boiling Pt: 121.1 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1 III
Merck Index: 12,09332
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test
Boiling point 120 - 122 °C
Density (20/4) 1.620 - 1.630
Free acidity Max. 0.0003 meq/g
Refractive index (20°C) 1.503 - 1.505
Evaporation residue Max. 10 ppm
Free chlorine Max. 2 ppm
Water Max. 50 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR