trans-4-Stilbenemethanol

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: 4-Hydroxymethylstilbene

262013-1G
262013-1GEA 119 GBP
262013-1G
trans-4-Stilbenemethanol
trans-4-Stilbenemethanol
Formula: C₁₅H₁₄O
MW: 210.27 g/mol
MDL Number: MFCD00011548
CAS Number: 101093-37-2

Order Now

Specification Test Results

Appearance (Colour) White to Yellow
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Purity (GC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR