Chlorophosphonazo III for spectrophotometric detection of alkaline earth metals, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

26049-1G-F
26049-1G-FEA 727 GBP
26049-1G-F
Chlorophosphonazo III for spectrophotometric detection of alkaline earth metals, Supelco®
Chlorophosphonazo III

Chlorophosphonazo III is a highly sensitive reagent and can be used for the spectrophotometric determination of zirconium, titanium, thorium and scandium


Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.

Formula: C₂₂H₁₆Cl₂N₄O₁₄P₂S₂
MW: 757.36 g/mol
MDL Number: MFCD00042698
CAS Number: 1914-99-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=050164d48875caa9ac8bff1ae5d16abc05a58381&vid=36511dbb28a6b762f712af6ec2f902ebd6949135&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR