Nickel(II) chloride hexahydrate 97, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Nickel dichloride hexahydrate

25850.290EA 231 GBP
25850.290
Nickel(II) chloride hexahydrate 97, GPR RECTAPUR®
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 140
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,06529

Order Now

Specification Test Results

Assay Min. 97 %
SO4 (Sulphate) Max. 0.1 %
Cd (Cadmium) Max. 20 ppm
Cu (Copper) Max. 20 ppm
Fe (Iron) Max. 100 ppm
Pb (Lead) Max. 20 ppm
Zn (Zinc) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR