Magnesium oxide heavy, TECHNICAL

Supplier: VWR Chemicals

Synonyms: Magnesia usta

25056.460EA 690 GBP
25056.460
Magnesium oxide heavy, TECHNICAL
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800 °C
Density: 3.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Assay (on ignited basis) Min. 85%
Loss on ignition (800°C) Max. 8%
Silica 3 - 6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR