Magnesium sulphate heptahydrate USP, J.T.Baker®

Supplier: Avantor

Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

M.O.C. Category R

2505-07EA 1330 GBP
2505-07
Magnesium sulphate heptahydrate USP, J.T.Baker®
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification Passes Test
USP - pH (1 in 20) 5.0 - 9.2
USP - Loss on Ignition 40.0 - 52.0 %
USP - Chloride (Cl) ≤ 0.014 %
USP - Iron (Fe) ≤ 0.5 ppm
USP - Selenium (Se) ≤ 0.003 %
USP - Assay (MgSO₄) (anhydrous basis) 99.0 - 100.5 %
USP - Endotoxin Concentration (2.5 EU/g max.) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
This product is intended for use in preparing parenteral and
nonparenteral dosage forms.
Elemental Impurities (USP 232, EP 5.20) - Information on
elemental impurities for this product is available on the
associated Product Regulatory Data Sheet and elemental impurity
profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR