Lithium chloride 98, purified

Supplier: VWR Chemicals
Warning

25009.236EA 59.5 GBP
25009.236 25009.293 25009.362 25009.464
Lithium chloride 98, purified
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR