n-Heptane 99, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: high purity solvents

24549.292EA 63.4 GBP
24549.292 24549.326 24549.361 24549.463 24549.554 24549.950
n-Heptane 99, GPR RECTAPUR®
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90,6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
REACH: 01-2119457603-38
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test
Boiling point 98 - 99 °C
Density (20/4) 0.681 - 0.685
Acidity Max. 0.0008 meq/g
Substances coloured by H2SO4 Max. 90 APHA
Evaporation residue Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR