VWR® Clamps, Three Pronged
Supplier: VWR Collection
241-0019EA 255 GBP
241-0019
VWR® Clamps, Three Pronged
Clamps General Purpose Clamps
Laboral aluminium alloy.
  • Two easy action knurled screws for adjusting the clamping fingers independently of each other