Petroleum spirit, 100…120 °C, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

23845.323EA 92.9 GBP
23845.323 23845.460 23845.551
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Petroleum spirit
Boiling Pt: 100…120 °C (1013 hPa)
Density: 0.7 g/cm³ (20 °C)
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

Substances coloured by H₂SO₄ Passes test
Acidity Max. 0.0003 meq/g
Density (20/4) 0.690 - 0.750
Distillation (min. 90 %) 100 - 125 °C
Evaporation residue Max. 20 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR