Diethyl ether 99 Stabilised, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilisation: 4-7 ppm BHT

23809.294EA 47.5 GBP
23809.294 23809.328 23809.363 23809.465
Diethyl ether 99 Stabilised, GPR RECTAPUR®
Diethyl ether
Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Assay (Ionol/BHT) 4 - 10 ppm
Acidity Max. 0.0003 meq/g
Evaporation residue Max. 20 ppm
Water Max. 0.2 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR