Disposable scalpels, sterile
Supplier: SWANN MORTON
0501 0503 0505 0506 0507 0508 0510 0511 0502 0518 0519 0520 0521 0522 0523 0524 0509 0516 0517 0504 0592 0591 0525 0514 0513 0512
233-5363EA 19.5 GBP
233-5363 233-5364 233-5366 233-5524 233-5525 233-5526 233-5528 233-5529 233-5513 233-5516 233-5517 233-5519 233-5520 233-5521 233-5522 233-5523 233-5527 233-0034 233-0035 233-5365 233-0154 233-0153 233-0152 233-0109 233-0206 233-0205
Disposable scalpels, sterile
Scalpels and Scalpel Assemblies Scalpels
Stainless steel blade with plastic handle.
  • Ergonomically-shaped handle
  • Clean, smooth cut with precision-ground blade
Packaging: Individually wrapped, sterile.
Order Now
SPECIFICATIONS
Environmentally Preferable