Cyclohexane 99, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

23223.290EA 57.8 GBP
23223.290 23223.324 23223.368 23223.461 23223.552
Cyclohexane 99, GPR RECTAPUR®
Cyclohexane
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 7
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
REACH: 01-2119463273-41
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test
Substances coloured by H2SO4 Passes test
Boiling point 79 - 81 °C
Density (20/4) 0.778 - 0.779
Free acidity Max. 0.0002 meq/g
Solidification point 5.5 - 6.5 °C
Evaporation residue Max. 50 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR