Copper 99,5, turnings, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals

23055.232EA 65.4 GBP
23055.232 23055.298
Copper 99,5, turnings, AnalaR NORMAPUR® zA
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Assay Min. 99.5 %
Insolubility in nitric acid Max. 0.05 %
Ag (Silver) Max. 50 ppm
Fe + Al Max. 0.025 %
Pb (Lead) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR