Nickel diiodide, anhydrous ≥99.5% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Nickel (II) iodide, Nickel iodide (NiI2)

22893.06 22893.14
22893.06EA 50.2 GBP
22893.06 22893.14
Nickel diiodide, anhydrous ≥99.5% (metals basis)
Nickel diiodide

Purity: 99.5% CAS No.: 13462-90-3 Molecular Formula: NiI2 Formula Weight: 312.51 Physical Form: -8 Mesh Density: 5.834 Melting Point: 797° UN No.: 3077 MDL No.: MFCD00016264

Formula: I₂Ni
MW: 312.5 g/mol
Melting Pt: 797
Density: 5.834
Storage Temperature: Ambient
MDL Number: MFCD00016264
CAS Number: 13462-90-3
EINECS: 236-666-6
UN: 3077
ADR: 9 III
Merck Index: 12,06594

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR