Tubing, rubber, vacuum
Supplier: DEUTSCH NEUMANN
3020515 3020616 3020818 3021020 3020717 3020412 3021026 3020818A 3021222 3020820 3021228 3021531 3020309 3020513 3020715 3020824 3020614 3020618 3020816 3021016 3020412A 3020612 3021030 3020816A
189-9122EA 209 GBP
189-9122 189-9123 189-9125 189-9126 189-9127 189-9128 189-9129 189-9130 228-1375 228-1551 228-1555 228-1556 224-0176 228-3897 228-3898 228-3899 228-1547 228-1548 228-1549 228-1552 228-1545 228-1546 228-1553 228-1550
Tubing, rubber, vacuum
Tubing Transfer Tubing
Natural rubber, red.
  • Flexible, excellent tensile strength
  • Economical
Heavy wall, suitable for vacuum applications in the laboratory.
Temperature -resistance: −10 to +50 °C.
Hardness (Shore durometer A): 50.
Order Now
SPECIFICATIONS
Environmentally Preferable