Pump tubing Tygon® LMT 55
Supplier: Saint Gobain Life Sciences
070534-02-ND 070534-03-ND 070534-11L-ND 070534-05L-ND 070534-08L-ND 070534-15L-ND 070534-17L-ND 070534-20L-ND 070534-21L-ND 070534-22L-ND 070534-24L-ND 070534-25L-ND 070534-23L-ND 070534-05-ND 070534-06-ND 070534-07-ND 070534-08-ND 070534-09-ND 070534-11-ND 070534-13-ND 070534-15-ND 070534-16-ND 070534-17-ND 070534-18-ND 070534-19-ND 070534-20-ND 070534-21-ND 070534-22-ND 070534-23-ND 070534-24-ND 070534-25-ND 070535-05-ND 070535-08-ND 070535-09-ND 070535-10-ND 070535-11-ND 070535-13-ND 070535-14-ND 070535-20-ND 070535-16-ND 070535-17-ND 070535-21-ND 070535-23-ND 070535-24-ND 070535-25-ND
224-0145EA 106 GBP
224-0145 VERN070534-03-ND VERN070534-11L-ND 228-1686 VERN070534-08L-ND VERN070534-15L-ND VERN070534-17L-ND VERN070534-20L-ND 224-0152 VERN070534-22L-ND 224-0154 VERN070534-25L-ND VERN070534-23L-ND VERN070534-05-ND VERN070534-06-ND VERN070534-07-ND VERN070534-08-ND 224-0147 VERN070534-11-ND VERN070534-13-ND VERN070534-15-ND 224-0149 VERN070534-17-ND VERN070534-18-ND VERN070534-19-ND VERN070534-20-ND 224-0151 VERN070534-22-ND VERN070534-23-ND 224-0153 VERN070534-25-ND VERN070535-05-ND VERN070535-08-ND VERN070535-09-ND VERN070535-10-ND VERN070535-11-ND VERN070535-13-ND VERN070535-14-ND VERN070535-20-ND VERN070535-16-ND 224-2012 VERN070535-21-ND VERN070535-23-ND VERN070535-24-ND VERN070535-25-ND
Pump tubing Tygon® LMT 55
Tubing Transfer Tubing
Non DEHP and phthalate-free for laboratory applications.
  • Outstanding chemical resistance
  • Lot-to-Lot consistency for reproducible results
  • Contains no BPA or phthalates
Crystal clear and flexible, long lasting and crack resistant, the non DEHP tubing delivers the same superior performance you have come to expect, but now in a formulation that contains a non DEHP plasticiser. Compatible with most inorganic chemicals found in the laboratory. It is non resistance to flex-fatigue and abrasioxidising and non contaminating, and less permeable than rubberMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable