VWR®, Funnels with Long Stem
Supplier: VWR Collection
221-0172EA 32.9 GBP
221-0172 221-0173 221-0174
VWR®, Funnels with Long Stem
Funnels Filtration Funnels Filter Funnels
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Smooth internal walls.
Certifications: ISO 4798.
Order Now
SPECIFICATIONS
Environmentally Preferable