VWR®, Stoppers for Standard Ground Joints
Supplier: VWR Collection
217-0210EA 13.3 GBP
217-0210 217-0211 217-0212 217-0213 217-0214 217-0215
VWR®, Stoppers for Standard Ground Joints
Stoppers PE/LDPE/HDPE Stoppers
Greaseless closure, which prevents ground joints from seizing. With inclined, circumferential sealing lips for better sealing and reduced risk of breakage. Easily detachable. With ergonomically shaped grip.
  • PTFE, solid
  • Excellent chemical resistance
  • Gas, liquid and vacuum-tight
  • Temperature resistant from –200 to +300 °C
Order Now
SPECIFICATIONS
Environmentally Preferable