Azomethine H sodium salt hydrate zA

Supplier: VWR Chemicals

Synonyms: Sodium 5-hydroxy-4-((2-hydroxybenzylidene)amino)-7-sulphonaphthalene-2-sulphonate xhydrate, Azomethine-H monosodium salt hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate, Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate, Azomethine-H sodium hydrate

21645.124EA 120 GBP
21645.124
Azomethine H sodium salt hydrate zA
Azomethine H sodium salt hydrate
Formula: C₁₇H₁₂NNaO₈S₂·xH₂O
MW: 463.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149588
CAS Number: 206752-32-1
EINECS: 227-698-1

Order Now

Specification Test Results

Suited for boron spectrophotometry Passes test
Water Max. 8 %
A (Lambda max.) (1 %; buffer pH 5.0) Min. 1100
Absorption maximum 231 - 241 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR