Carboys, wide neck, rectangular with spigot, HDPE, Nalgene®
Supplier: Thermo Fisher Scientific
2320-0020 2320-0050 2321-0020 2321-0050 2322-0020 2322-0050
216-8611EA 111 GBP
216-8611 216-8612 216-8631 216-8632 216-8651 216-8652
Carboys, wide neck, rectangular with spigot, HDPE, Nalgene®
Bottles Carboys
HDPE, translucent, PPCO, translucent or PC, transparent.
  • HDPE: Good chemical resistance/PPCO: Very good chemical resistance/PC: Good chemical resistance
  • HDPE: Heat resistant up to 120 °C/PPCO: Heat resistant up to 121 °C, autoclavable/PC: Heat resistant up to 135 °C, autoclavable
  • Leakproof closure
With white PP screw caps and PP spigot. Space saving rectangular shape. Calibrated in gallons and litres. Wide opening for easy filling and decanting or cleaning.  Sturdy stainless steel handle. The carboy made of PC (heat resistant up to 135 °More Product Information