Jars, wide mouth, with screw cap
Supplier: KARTELL
1563 1564 1565 1566 1567 1568 1569
216-2403EA 383 GBP
216-2403 216-2404 216-2405 216-2406 216-2407 216-2408 216-5742
Jars, wide mouth, with screw cap
Jars General Use Jars
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from –50 to +80 °C
  • Wide opening makes it ideally suited for storing and transporting powders and pastes
With black screw cap and sealing element.
Certifications: Suitable for foodstuff according to EC regulation 10/2011.