Sample containers with screw cap, Bijou, WHEATON®
Supplier: DWK Life Sciences
327145 327147 327149
216-0979EA 209 GBP
216-0979 216-0980 216-0981
Sample containers with screw cap, Bijou, WHEATON®
Sample Containers General Purpose Containers
PS clear, with PE screw cap.
  • Manufactured from FDA approved virgin PS
  • Supplied sterile and for in vitro use only
  • PS offers good thermal and chemical resistance
  • All materials are non-cytotoxic
Straight-walled vessels ideal for sample collection, storage and transport. Supplied with leak proof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certifications: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
Packaging: Packed in inner bags of 50.
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable