Bottles, amber, narrow neck, round, HDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
2004-0001 2004-0002 2004-0004 2004-0008 2004-0016 2004-0032 2004-9025 2004-9050 2004-9125
215-7544EA 19 GBP
215-7544 215-7545 215-7546 215-7548 215-7550 215-7551 215-7542 215-7543 215-7541
Bottles, amber, narrow neck, round, HDPE, with screw caps, Nalgene®
Bottles Plastic Bottles
HDPE, amber with amber PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 120 °C
  • Leakproof
Amber to protect light-sensitive materials. Ideal for storing and shipping liquids.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement
NAR Certificate of Retirement ISO14001 (Certificate Registration No. EMS 752603)