VWR® Scintillation Vials
Supplier: VWR Collection
215-4488EA 101 GBP
215-4488 215-4491
VWR® Scintillation Vials
Vials
VWR® scintillation vials are produced from borosilicate 3.3 glass. They are compatible with all scintillation counters.
  • High resistance against chemicals and thermal shock
  • Low background counts and high ultraviolet transmission
  • Delivered in convenient pack sizes
VWR® scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting or for sample storage.
Ordering information: Screw caps have to be ordered separately and you can choose the right screw cap for your application from our selection of fitting screw caps withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable