Containers, wide mouth, square with thread
Supplier: Kautex
310-70597 310-70598 310-70599 310-70699 310-70700 310-70701 310-71080 310-71081 310-71082 310-71083 310-73352 310-73353 310-70596 310-70698 310-70595 310-70697
215-3906EA 1.3 GBP
215-3906 215-3908 215-3930 215-3916 215-3918 215-3932 215-3907 215-3917 215-3910 215-3920 215-3931 215-3933 215-3116 215-3118 215-2185 215-2186
Containers, wide mouth, square with thread
Sample Containers General Purpose Containers
PVC, square, transparent or brown transparent colours.
  • Wide neck opening
  • Easy to fill, easy to clean
  • Leakproof
  • Suitable for food
  • Halal certified
Due to the wide neck opening the containers are easy to fill and to clean. Black closure with PE-foam insert. Brown transparent containers are suitable for light-sensitive media.
Order Now
SPECIFICATIONS
Environmentally Preferable