VERAL Bottles, Clear or Brown Glass, with Thread
Supplier: RIXIUS
1-0401-0050-28 1-0400-0500-28 1-0400-1000-28 1-0400-0100-28 1-0400-0200-28 1-0400-0250-28 1-0400005028PP 1-0401-0250-28 1-0401-0060-28 1-0401-0150-28 1-0401-0100-28 1-0401-0500-28 1-0401-1000-28 1-0401003028AD 1-0400-0030-28 1-0401-0200-28
RIXI1-0401-0050-28EA 243 GBP
RIXI1-0401-0050-28 215-2859 215-2860 215-2855 215-2857 215-2858 215-2854 215-2866 215-2862 215-2864 215-2863 215-2867 215-2868 215-3606 RIXI1-0400-0030-28 215-4420
VERAL Bottles, Clear or Brown Glass, with Thread
Bottles Glass Bottles
Clear or brown glass, with thread.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Closures to ordered separately
Light packaging bottles. Excellent for sampling, storing and transporting liquids. Brown coloured to protect light-sensitive materials.
Order Now
SPECIFICATIONS
Environmentally Preferable