Laboratory bottles, narrow neck, with thread, DURAN®
Supplier: DWK Life Sciences
218011404 218011704 218012409 218013602 218014401 218015406 218016308 218017304 218018609 218019108 218018806 218012906 218015106 218016908 218010802
215-1532EA 8.7 GBP
215-1532 215-1533 215-1534 215-1535 215-1536 215-1537 215-1538 215-1539 215-1540 215-1542 215-1541 215-2425 215-2426 215-2427 215-3295
Laboratory bottles, narrow neck, with thread, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear, without screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Round, with graduation and DIN thread. Without pouring ring or screw cap.
With printed retrace code (batch identification).
*With specially shaped glass rim for improved pouring; no additional pouring ring is needed.
**Ring-shaped reinforcement on the bottle shoulder makes the filling line of the nominalMore Product Information