Bottles, narrow neck, with standard ground stoppers, DURAN®
Supplier: DWK Life Sciences
211651702 211652407 211653609 211654408 211655404 211656306 211657302 211658607 211659106 211681705 211682401 211683603 211684402 211685407 211686309 211687305 211688601 211689109
215-1113EA 41.9 GBP
215-1113 215-1114 215-1115 215-1116 215-1117 215-1118 215-1119 215-1120 215-1121 215-1143 215-1144 215-1145 215-1146 215-1147 215-1148 215-1149 215-1150 215-1151
Bottles, narrow neck, with standard ground stoppers, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass type I according to USP, EP and JP
  • Amber glass meets USP <671> and Eur. Ph. 3.2.1. for protection against UV light
Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
ISO 4796-2.
With printed Retrace Code (batch identification).
Order Now
SPECIFICATIONS
Environmentally Preferable