Bottles, wide neck, square, with screw caps, Azlon®
Supplier: SciLabware
BGE430P
215-0460EA 141 GBP
215-0460
Bottles, wide neck, square, with screw caps, Azlon®
Bottles Plastic Bottles
HDPE, translucent with black wadded screw caps.
  • Good chemical resistance
  • Heat-resistant up to 120 °C
  • Economical on shelf space
Ideal for storage of solids. Wide opening allows rapid filling and dispensing of powders, pastes and granules. Practical rounded square design.