Methyl decanoate, Supelco®

Supplier: Merck

Synonyms: Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate

MQ100

21479-1ML 21479-5ML
21479-1MLEA 60 GBP
21479-1ML 21479-5ML
Methyl decanoate, Supelco®
Methyl decanoate
Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 224
Melting Pt: –18 °C
Density: 0.8724 g/cm³ (20 °C)
Flash Pt: 94
MDL Number: MFCD00009580
CAS Number: 110-42-9
EINECS: 203-766-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12ccd05fc8b1d288c4644ba8b4dddd40fa093561&vid=96a4236d4b7d184a4ef1271008c8eb705aff34c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR