Silica gel, Davisil® Grade 12 pore size 22 Å, 28-200 mesh, high_purity, Supelco®

Supplier: Merck

214396-1KG 214396-5KG
214396-1KGEA 125 GBP
214396-1KG 214396-5KG
Silica gel, Davisil® Grade 12 pore size 22 Å, 28-200 mesh, high_purity, Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C
Melting Pt: > 1600 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeb910a95beceac796ecc2e40cfbae6327b16611&vid=39912bddd50790ed76c141635a89325146f5ab7d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR