Ammonium sulphide 20% in aqueous solution, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

21352.292EA 214 GBP
21352.292 21352.441
Ammonium sulphide 20% in aqueous solution, GPR RECTAPUR®
Ammonium sulphide
Formula: H₈N₂S
MW: 68.14 g/mol
Melting Pt: -18
Density: 0.99
MDL Number: MFCD00010892
CAS Number: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8 (3, 6.1) II
Merck Index: 13,00560

Order Now

Specification Test Results

Assay (as (NH4)2S from sulphide) 17 - 23 %
Ignition residue (SO4) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR